Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145965.2 | NP_666077.2 | 4, 5 | 425 | ||
NM_172952.3 | NP_766540.2 | 4, 5 | 425 | |||
GenBank mRNA | AK082353.1 | - | 4, 5 | 347 | ||
AK144569.1 | - | 4, 5 | 428 | |||
BC127058.1 | - | 4, 5 | 347 | 40109771 |