Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166033.1 | NP_001159505.1 | 3 | 625 | ||
NM_172977.3 | NP_766565.2 | 2 | 519 | |||
GenBank mRNA | AK030351.1 | - | 2 | 446 | ||
AK141269.1 | - | 2 | 500 | |||
AK146401.1 | - | 3 | 581 | |||
AK146403.1 | - | 3 | 581 | |||
AK150650.1 | - | 2 | 501 | |||
AK151179.1 | - | 2 | 501 | |||
AK153102.1 | - | 2 | 501 | |||
BC061476.1 | - | 2 | 452 | |||
BC095998.1 | - | 2 | 458 | 30736671 | ||
BC117919.1 | - | 2 | 431 | 40097451 | ||
BC117920.1 | - | 2 | 429 | 40097456 | ||
BU610856.1 | - | 3 | 552 | |||
CN530016.1 | - | 2 | 519 |