Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109902.1 | NP_001103372.1 | 4 | 719 | ||
NM_172998.3 | NP_766586.2 | 5 | 754 | |||
GenBank mRNA | AK046850.1 | - | 5 | 755 | ||
AK090256.1 | - | 4 | 815 | |||
AK148930.1 | - | 4 | 696 | |||
BC057302.1 | - | 4 | 681 | 6827287 |