Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001153.4 | NP_001144.1 | 6 | 580 | ||
NM_001320698.1 | NP_001307627.1 | 8 | 1862 | |||
NM_001320700.1 | NP_001307629.1 | 6 | 514 | |||
NM_001320702.1 | NP_001307631.1 | 6 | 615 | |||
XM_017003943.1 | XP_016859432.1 | 9 | 1724 | |||
XM_017003944.1 | XP_016859433.1 | 9 | 1690 | |||
GenBank mRNA | AB209457.1 | - | 6 | 459 | ||
AK058017.1 | - | 6 | 420 | |||
AK293177.1 | - | 6 | 487 | |||
AK293396.1 | - | 6 | 352 | |||
AK293406.1 | - | 6 | 368 | |||
AK303960.1 | - | 6 | 356 | |||
AK315511.1 | - | 6 | 420 | |||
AY453398.1 | - | 5 | 364 | |||
BC000182.1 | - | 6 | 462 | 3352245 | ||
BC011659.1 | - | 6 | 440 | 4301675 | ||
BC063672.1 | - | 6 | 514 | 3908264 | ||
BE886769.1 | - | 493 | ||||
CR407681.1 | - | 4 | 332 | |||
D78152.1 | - | 5 | 394 | |||
FJ224287.1 | - | 6 | 405 | |||
M19383.1 | - | 6 | 405 | |||
M82809.1 | - | 6 | 405 |