Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172924.3 | NP_766512.2 | 5 | 3690 | ||
GenBank mRNA | AK048912.1 | - | 5 | 3677 | ||
AK090133.1 | - | 5 | 3704 | |||
AK173328.1 | - | 2 | 232 |