Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290786.1 | NP_001277715.1 | 3 | 959 | ||
NM_172979.3 | NP_766567.1 | 3 | 959 | |||
GenBank mRNA | AK028832.1 | - | 2 | 154 | ||
AK052840.1 | - | 3 | 846 | |||
AK142590.1 | - | 3 | 846 | |||
AY198336.1 | - | 3 | 877 | |||
AY198337.1 | - | 3 | 844 | |||
BC055469.1 | - | 2 | 127 | |||
BC094506.1 | - | 3 | 842 | 5341608 | ||
BC116796.1 | - | 3 | 838 | 40126115 | ||
BC119176.1 | - | 3 | 838 | 8733925 |