Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173030.2 | NP_766618.2 | 10 | 1725 | ||
GenBank mRNA | AB082928.1 | - | 8 | 1213 | ||
AK030726.1 | - | 9 | 1657 | |||
AK038387.1 | - | 10 | 1726 | |||
AK045417.1 | - | 10 | 1726 | |||
BC131651.1 | - | 10 | 1404 | 40110517 | ||
BC131652.1 | - | 9 | 1435 | 40110518 |