Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199283.1 | NP_001186212.1 | 2 | 481 | ||
NM_001199284.1 | NP_001186213.1 | 2 | 263 | |||
NM_173388.2 | NP_775564.1 | 2 | 226 | |||
GenBank mRNA | AB120363.1 | - | 2 | 259 | ||
AK090207.1 | - | 1 | 560 | |||
AK143773.1 | - | 2 | 324 | |||
AK159144.1 | - | 2 | 449 | |||
AK165721.1 | - | 1 | 596 | |||
AK171104.1 | - | 2 | 481 | |||
AK172685.1 | - | 2 | 251 | |||
BC042513.1 | - | 2 | 222 | 3964020 | ||
CD550550.1 | - | 2 | 222 |