Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173404.3 | NP_775580.1 | 1 | 847 | ||
GenBank mRNA | AK033644.1 | - | 1 | 485 | ||
AK035299.1 | - | 1 | 873 | |||
AK035393.1 | - | 1 | 836 | |||
AK037225.1 | - | 1 | 828 | |||
AK040389.1 | - | 1 | 848 | |||
BC117749.2 | - | 1 | 460 | 40090489 |