Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173748.4 | NP_776109.1 | 6 | 3678 | ||
GenBank mRNA | AK049739.1 | - | 6 | 3390 |