Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001236.3 | NP_001227.1 | 3 | 706 | ||
GenBank mRNA | AB004850.1 | - | 2 | 137 | ||
AB004854.1 | - | 3 | 478 | |||
AB041012.1 | - | 3 | 759 | |||
AB124847.1 | - | 3 | 653 | |||
AK223429.1 | - | 3 | 584 | |||
BC002812.2 | - | 3 | 566 | 3637434 | ||
CR541709.1 | - | 3 | 478 | |||
FJ224313.1 | - | 3 | 478 |