Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304743.1 | NP_001291672.1 | 18 | 5276 | ||
NM_014720.3 | NP_055535.2 | 19 | 5369 | |||
XM_011540401.2 | XP_011538703.1 | 18 | 4025 | |||
GenBank mRNA | AB002804.1 | - | 18 | 4949 | ||
AF273048.1 | - | 19 | 4842 | |||
BX648375.1 | - | 1 | 2673 | |||
D86959.1 | - | 18 | 5224 |