Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173762.4 | NP_776123.3 | 36 | 5621 | ||
GenBank mRNA | AK049676.1 | - | 3 | 423 | ||
AY493378.1 | - | 36 | 5534 | |||
BC049989.1 | - | 2 | 186 | |||
BC059032.1 | - | 4 | 583 |