Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1nb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166619.1 | NP_001160091.1 | 4 | 591 | ||
NM_001166620.1 | NP_001160092.1 | 3 | 513 | |||
NM_174993.2 | NP_778158.2 | 5 | 729 | |||
GenBank mRNA | AK076190.1 | - | 4 | 549 | ||
AY214130.1 | - | 5 | 634 | |||
BC099411.1 | - | 4 | 591 | 30788714 | ||
BC138087.1 | - | 4 | 572 | 8861106 | ||
BC145139.1 | - | 5 | 710 | 9053689 | ||
BC145140.1 | - | 5 | 710 | 9053690 | ||
BC171922.1 | - | 3 | 494 | 9053688 | ||
BC171923.1 | - | 4 | 572 | 9053691 | ||
BC171947.1 | - | 5 | 710 | 9054233 | ||
BC171948.1 | - | 5 | 710 | 9054234 |