Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175091.3 | NP_780300.2 | 21 | 3255 | ||
GenBank mRNA | AK048860.1 | - | 17 | 2620 | ||
AK163725.1 | - | 1 | 82 | |||
BC057370.1 | - | 21 | 3236 | 6835784 |