Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289545.1 | NP_001276474.1 | 4 | 482 | ||
NM_175183.5 | NP_780392.1 | 5 | 661 | |||
GenBank mRNA | AK046849.1 | - | 5 | 1069 | ||
BC027136.1 | - | 2 | 445 | |||
BC088996.1 | - | 3 | 256 | |||
BC094547.1 | - | 4 | 417 | |||
BC137599.1 | - | 4 | 403 | 8860618 | ||
BC137600.1 | - | 4 | 430 | 8860619 | ||
BC144756.1 | - | 4 | 410 | 9053304 |