Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006967.1 | - | 2 | 711 | ||
AK088316.1 | - | 4 | 1158 | |||
AK164311.1 | - | 2 | 393 |