Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114.4 | NP_001105.1 | 25 | 4738 | ||
XM_011522835.2 | XP_011521137.1 | 26 | 4857 | |||
XM_011522836.2 | XP_011521138.1 | 26 | 4873 | |||
XM_011522837.2 | XP_011521139.1 | 26 | 4802 | |||
XM_011522838.2 | XP_011521140.1 | 26 | 5006 | |||
XM_011522839.2 | XP_011521141.1 | 26 | 4833 | |||
XM_011522840.2 | XP_011521142.1 | 26 | 4840 | |||
XM_011522842.2 | XP_011521144.1 | 26 | 4903 | |||
XM_017022895.1 | XP_016878384.1 | 26 | 4855 | |||
XM_017022896.1 | XP_016878385.1 | 23 | 4019 | |||
XM_017022897.1 | XP_016878386.1 | 23 | 4038 | |||
GenBank mRNA | BC126271.1 | - | 25 | 4549 | 8991987 | |
D25538.1 | - | 25 | 4735 |