Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pogk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142948.1 | NP_001136420.1 | 2 | 186 | ||
NM_175170.4 | NP_780379.2 | 3 | 520 | |||
GenBank mRNA | AK031716.1 | - | 3 | 473 | ||
AK048067.1 | - | 3 | 479 | |||
AK048752.1 | - | 2 | 145 | |||
AK053898.1 | - | 3 | 497 | |||
AK122520.1 | - | 2 | 458 | |||
BC072587.1 | - | 140 | 6829796 |