Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126.3 | NP_001117.2 | 4 | 702 | ||
XM_011544108.2 | XP_011542410.1 | 4 | 1374 | |||
GenBank mRNA | AK025514.1 | - | 4 | 449 | ||
AK303886.1 | - | 5 | 414 | |||
AK308749.1 | - | 5 | 787 | |||
BC012356.1 | - | 4 | 487 | 4635347 |