Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MELK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256685.1 | NP_001243614.1 | 5 | 466 | ||
NM_001256689.1 | NP_001243618.1 | 4 | 380 | |||
NM_001256690.1 | NP_001243619.1 | 3 | 263 | |||
NM_001256693.1 | NP_001243622.1 | 3 | 284 | |||
NM_014791.3 | NP_055606.1 | 5 | 466 | |||
NR_046337.1 | 4 | 380 | ||||
XM_011518076.2 | XP_011516378.1 | 5 | 645 | |||
XM_011518077.1 | XP_011516379.1 | 6 | 541 | |||
XM_011518078.2 | XP_011516380.1 | 6 | 728 | |||
XM_011518079.1 | XP_011516381.1 | 5 | 418 | |||
XM_011518080.1 | XP_011516382.1 | 5 | 433 | |||
XM_011518081.2 | XP_011516383.1 | 5 | 451 | |||
XM_011518082.2 | XP_011516384.1 | 4 | 559 | |||
XM_011518083.2 | XP_011516385.1 | 5 | 642 | |||
XM_011518084.2 | XP_011516386.1 | 4 | 254 | |||
XM_011518085.1 | XP_011516387.1 | 3 | 250 | |||
XM_011518086.2 | XP_011516388.1 | 5 | 433 | |||
GenBank mRNA | AB183427.1 | - | 4 | 331 | ||
AB183428.1 | - | 3 | 216 | |||
AK222811.1 | - | 5 | 465 | |||
AK293447.1 | - | 3 | 220 | |||
AK296820.1 | - | 4 | 379 | |||
AK299164.1 | - | 5 | 418 | |||
AK300821.1 | - | 4 | 334 | |||
AK301131.1 | - | 3 | 216 | |||
AK309470.1 | - | 3 | 241 | |||
BC014039.2 | - | 5 | 420 | 4547136 | ||
BP365611.1 | - | 5 | 468 | |||
D79997.1 | - | 5 | 452 |