Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175199.3 | NP_780408.1 | 3 | 331 | ||
GenBank mRNA | AB093239.1 | - | 3 | 275 | ||
AK006867.1 | - | 3 | 315 | |||
AK038491.1 | - | 3 | 303 | |||
AK087732.1 | - | 4 | 488 | |||
AK141925.1 | - | 3 | 332 | |||
AK165073.1 | - | 3 | 359 | |||
AY196789.1 | - | 3 | 330 | |||
BC030362.1 | - | 3 | 301 | 5369051 |