Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028785.3 | NP_083061.2 | 16 | 1969 | ||
GenBank mRNA | AK038285.1 | - | 15 | 1717 | ||
BC141358.1 | - | 16 | 1964 | 9055970 |