Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175314.3 | NP_780523.2 | 15 | 2094 | ||
GenBank mRNA | AK030699.1 | - | 2 | 204 | ||
AK173146.1 | - | 2 | 214 | |||
BC068142.1 | - | 1 | 133 | |||
CK793536.1 | - | 5 | 626 |