Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081057.2 | NP_001074526.1 | 21 | 7409 | ||
NM_001289510.1 | NP_001276439.1 | 20 | 7357 | |||
NM_175336.3 | NP_780545.2 | 21 | 7519 | |||
GenBank mRNA | AK029810.1 | - | 3 | 3334 | ||
AK129113.1 | - | 20 | 7362 | |||
AK147592.1 | - | 20 | 7358 |