Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930555G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175393.4 | NP_780602.2 | 1 | 190 | ||
GenBank mRNA | AK076920.1 | - | 1 | 191 |