Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252372.1 | NP_001239301.1 | 24 | 3187 | ||
NM_175418.5 | NP_780627.2 | 25 | 3229 | |||
GenBank mRNA | AK078761.1 | - | 5 | 623 | ||
AK132188.1 | - | 24 | 3060 | |||
BC062158.1 | - | 25 | 3091 | 30134842 |