Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003717.1 | NP_001003717.1 | 3, 4 | 646 | ||
NM_175489.3 | NP_780698.2 | 4, 5 | 683 | |||
GenBank mRNA | AK037158.1 | - | 4, 5 | 683 | ||
AK050244.1 | - | 3, 4 | 647 | |||
BC141383.1 | - | 4, 5 | 382 | 9055996 |