Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A930002I21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080695.1 | - | 2 | 642 | ||
AK088812.1 | - | 2 | 724 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_175410.1 | - | 642 | Chr. - - [-] on Build |