Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175472.3 | NP_780681.2 | 6 | 1688 | ||
GenBank mRNA | AK031299.1 | - | 4 | 1081 | ||
AK049279.1 | - | 4 | 677 | |||
AK220327.1 | - | 4 | 936 | |||
BC145507.1 | - | 5 | 1339 | |||
BC150791.1 | - | 5 | 1339 | 9087702 |