Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175391.4 | NP_780600.1 | 5 | 521 | ||
GenBank mRNA | AK075855.1 | - | 5 | 523 | ||
BC139148.1 | - | 5 | 480 | 8862170 | ||
BC139149.1 | - | 5 | 480 | 8862171 |
Gene Symbol: Apol7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164640.1 | NP_001158112.1 | 5 | 750 | ||
NM_029419.2 | NP_083695.2 | 6 | 850 | |||
GenBank mRNA | AK018646.1 | - | 6 | 852 | ||
AK085956.1 | - | 5 | 727 | |||
BC139137.1 | - | 5 | 629 | 8862159 | ||
BC139138.1 | - | 5 | 629 | 8862160 |
Gene Symbol: Apol7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024848.2 | NP_001020019.2 | 5 | 497 | ||
GenBank mRNA | BC085284.1 | - | 5 | 509 | 30750330 |