Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175406.3 | NP_780615.2 | 8 | 2209 | ||
GenBank mRNA | AB088358.1 | - | 8 | 2168 | ||
AK137586.1 | - | 8 | 2192 | |||
AK170405.1 | - | 8 | 2182 | |||
AK170578.1 | - | 8 | 2153 | |||
AY145896.1 | - | 8 | 2209 | |||
AY517482.1 | - | 8 | 2197 |