Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd34b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175455.4 | NP_780664.2 | 5 | 992 | ||
GenBank mRNA | AK078203.1 | - | 5 | 1066 | ||
AK090387.1 | - | 5 | 995 | |||
AK138465.1 | - | 4 | 1178 | |||
BC096609.1 | - | 5 | 868 | |||
BC119312.1 | - | 5 | 854 | 8734061 | ||
BC119314.1 | - | 5 | 854 | 8734063 |