Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175460.3 | NP_780669.1 | 11 | 2505 | ||
GenBank mRNA | AK083532.1 | - | 11 | 2608 | ||
AK084041.1 | - | 12 | 2715 | |||
AK129155.1 | - | 11 | 2588 | |||
BC062645.1 | - | 1 | 634 | |||
BC078443.1 | - | 1 | 314 | |||
BC089007.1 | - | 11 | 2505 | 30532596 |