Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175519.5 | NP_780728.3 | 1 | 548 | ||
GenBank mRNA | AK039167.1 | - | 1 | 518 | ||
AK043351.1 | - | 1 | 518 | |||
AK045439.1 | - | 1 | 401 | |||
AK047519.1 | - | 1 | 549 | |||
AK082563.1 | - | 1 | 402 | |||
AY615967.1 | - | 1 | 550 | |||
BC116940.1 | - | 1 | 199 | 40126259 | ||
BC116942.1 | - | 1 | 199 | 40126261 |