Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nim1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175538.3 | NP_780747.1 | 3 | 1145 | ||
GenBank mRNA | AK044462.1 | - | 2 | 850 | ||
AK047794.1 | - | 2, 3 | 1135 | |||
AK048694.1 | - | 3 | 1146 | |||
AK053734.1 | - | 3 | 1152 | |||
BC051189.1 | - | 2 | 812 | 6491737 | ||
BC106122.1 | - | 2 | 839 | 4505882 | ||
BC117955.1 | - | 3 | 1071 | 40098149 | ||
BC117956.1 | - | 3 | 1069 | 40098152 |