Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113360.2 | NP_001106831.1 | 2 | 419 | ||
NM_175556.4 | NP_780765.2 | 2 | 419 | |||
GenBank mRNA | AK044619.1 | - | 2 | 418 | ||
AK049970.1 | - | 2 | 263 | |||
AK078731.1 | - | 2 | 190 | |||
AK129147.1 | - | 2 | 119 | |||
AY966876.1 | - | 2 | 197 | |||
DQ176851.1 | - | 2 | 419 |