Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175510.3 | NP_780719.2 | 4, 5 | 601 | ||
GenBank mRNA | AK043914.1 | - | 4, 5 | 620 | ||
AK044008.1 | - | 4, 5 | 634 | |||
AK081235.1 | - | 4, 5 | 636 | |||
BC116429.1 | - | 4, 5 | 634 | |||
BC116430.2 | - | 4, 5 | 634 | 40104344 | ||
FJ210934.1 | - | 4, 5 | 601 |