Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031075.1 | - | 3 | 3663 | ||
AK038524.1 | - | 1 | 545 | |||
AK038736.1 | - | 1 | 545 |