Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195816.1 | NP_001182745.1 | 3 | 545 | ||
NM_176916.4 | NP_795890.2 | 3 | 849 | |||
GenBank mRNA | AK046294.1 | - | 3 | 812 | ||
AK082725.1 | - | 3 | 446 | |||
AK144116.1 | - | 3 | 838 | |||
BU609501.1 | - | 3 | 514 | |||
CK626039.1 | - | 3 | 545 |