Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110218.1 | NP_001103688.1 | 2 | 729 | ||
NM_176919.4 | NP_795893.2 | 2 | 729 | |||
GenBank mRNA | AK029461.1 | - | 2 | 291 | ||
AK040194.1 | - | 2 | 473 | |||
AK040207.1 | - | 2 | 714 | |||
AK043121.1 | - | 2 | 730 | |||
AK051434.1 | - | 2 | 687 | |||
AK077422.1 | - | 2 | 718 | |||
AK134804.1 | - | 2 | 709 | |||
AK150309.1 | - | 2 | 291 | |||
AK220260.1 | - | 2 | 686 | |||
BC052910.1 | - | 1 | 40 |