Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176920.4 | NP_795894.1 | 2 | 777 | ||
GenBank mRNA | AK044492.1 | - | 2 | 731 | ||
AK138618.1 | - | 2 | 595 | |||
BC099587.1 | - | 2 | 575 | 30289234 | ||
BC132594.1 | - | 2 | 481 | 40130871 | ||
BC145196.1 | - | 2 | 481 | |||
BC145990.1 | - | 2 | 481 | 40131293 |