Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Topbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176979.5 | NP_795953.2 | 4 | 290 | ||
GenBank mRNA | AK084654.1 | - | 4 | 393 | ||
AK085031.1 | - | 4 | 429 | |||
AK129104.1 | - | 4 | 402 | |||
BC049797.1 | - | 4 | 290 | 6510112 |