Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043389.1 | - | 2 | 1555 | ||
AK080683.1 | - | 2 | 1626 | |||
BC115977.1 | - | 1 | 1080 | 40105192 |