Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176940.5 | NP_795914.3 | 6 | 2106 | ||
GenBank mRNA | AK034868.1 | - | 1805 | |||
AK038792.1 | - | 5 | 1799 | |||
AK087622.1 | - | 6 | 2108 | |||
AK162888.1 | - | 6 | 2101 | |||
BC082552.1 | - | 6 | 2046 | |||
BC144786.1 | - | 4 | 1696 | 9053334 | ||
BC144787.1 | - | 4 | 1696 | |||
BC145703.1 | - | 4 | 1696 | 40131006 |