Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176980.4 | NP_795954.2 | 3 | 1148 | ||
GenBank mRNA | AK030065.1 | - | 3 | 1149 | ||
AK077036.1 | - | 2 | 528 | |||
BC132415.1 | - | 3 | 1123 | 40130692 | ||
BC145253.1 | - | 3 | 1123 | 9053805 |