Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A730020M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042746.1 | - | 2 | 228 | ||
AK085594.1 | - | 2 | 218 | |||
AK160476.1 | - | 2 | 340 | |||
BX522104.1 | - | 2 | 158 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_176992.1 | - | 227 | Chr. - - [-] on Build |