Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164086.1 | NP_001157558.1 | 9 | 9049 | ||
NM_001164087.1 | NP_001157559.1 | 9 | 9065 | |||
NM_011983.2 | NP_036113.1 | 9 | 9082 | |||
GenBank mRNA | AK033939.1 | - | 1974 | |||
AK085081.1 | - | 740 |