Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197295.1 | NP_001184224.1 | 2 | 284 | ||
NM_001197296.1 | NP_001184225.1 | 2 | 284 | |||
NM_001393.3 | NP_001384.1 | 2 | 284 | |||
XM_017014376.1 | XP_016869865.1 | 1 | 124 | |||
XM_017014377.1 | XP_016869866.1 | 1 | 124 | |||
GenBank mRNA | AB011792.1 | - | 2 | 170 | ||
AK296458.1 | - | 2 | 187 | |||
AK298278.1 | - | 3 | 196 | |||
AK303922.1 | - | 2 | 185 | |||
AK312820.1 | - | 2 | 182 | |||
BC105958.1 | - | 1 | 127 | 40035034 | ||
BC107493.1 | - | 1 | 127 | 40035036 | ||
BX537976.1 | - | 2 | 247 | |||
DC300418.1 | - | 2 | 285 |