Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036300.1 | - | 1 | 1037 |
Gene Symbol: B230354K17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045689.1 | - | 3 | 1483 | ||
AK046225.1 | - | 3 | 1483 | |||
AK083460.1 | - | 3 | 2427 | |||
AK171144.1 | - | 3 | 2427 | |||
BC023836.1 | - | 1 | 1630 | |||
BC027345.1 | - | 2 | 1630 |
Gene Symbol: Gm30289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035430.1 | - | 1 | 3405 |
Gene Symbol: Hmox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC055742.1 | - | 1 | 1999 |
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054315.1 | - | 2 | 1332 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK078717.1 | - | 769 | Chr. - - [-] on Build |